Learn More
Invitrogen™ TEAD2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total TEA domain family member 2.
Specifications
Specifications
| Antigen | TEAD2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TEAD2 |
| Gene Accession No. | P48301, Q15562 |
| Gene Alias | Embryonic TEA domain containing factor; embryonic TEA domain-containing factor; Etdf; ETEF-1; Etf; TEA domain family member 2; TEA domain transcription factor 2; Tead2; TEAD-2; TEF4; TEF-4; transcriptional enhancer factor TEF-4 |
| Gene Symbols | TEAD2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.