Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEAD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TEAD2 |
---|---|
Concentration | 0.29 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-100-2786
![]() ![]() |
Proteintech Group Inc
211591AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
TEAD2 is a putative transcription factor that binds to the SPH and GT-IIC enhansons (5′-GTGGAATGT-3′). TEAD2 may be involved in the gene regulation of neural development. TEAD2 binds to the M-CAT motif.Specifications
TEAD2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P48301, Q15562 | |
21677, 8463 | |
TEAD2 Fusion Protein Ag14074 | |
Primary | |
-20°C | |
TEAD2 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ETF, TEA domain family member 2, TEAD 2, TEAD2, TEF 4, TEF4 | |
TEAD2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title