Learn More
TEAD3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | TEAD3 |
|---|---|
| Concentration | 0.25 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8504
![]()
|
Proteintech Group Inc
131201AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This is a rabbit polyclonal anti-TEAD3 antibody raised against part of C-terminal chain of human TEAD3.
This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is predomitly expressed in the placenta and is involved in the transactivation of the chorionic somatomammotropin-B gene enhancer. Translation of this protein is initiated at a non-AUG (AUA) start codon.Specifications
| TEAD3 | |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P70210, Q99594 | |
| 21678, 7005 | |
| TEAD3 Fusion Protein Ag3875 | |
| Primary | |
| -20°C | |
| TEAD3 |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DTEF 1, ETFR 1, TEA domain family member 3, TEAD 3, TEAD3, TEAD5, TEF 5, TEF5 | |
| TEAD3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.