Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEAD4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220590
Description
TEAD4 Polyclonal specifically detects TEAD4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
TEAD4 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
EFTR-2, hRTEF-1B, related transcription enhancer factor 1B, RTEF1RTEF-1, TCF13L1MGC9014, TEA domain family member 4TEF3, TEAD-4, TEF-3, TEFR-1, Transcription factor 13-like 1, Transcription factor RTEF-1, transcriptional enhancer factor 1-related, transcriptional enhancer factor 3, transcriptional enhancer factor TEF-3 | |
Rabbit | |
48 kDa | |
0.1 mL | |
Stem Cell Markers | |
7004 | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
Q15561 | |
TEAD4 | |
Recombinant protein encompassing a sequence within the center region of human TEAD4. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (94%), Chicken (88%), Pig (96%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction