Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEAD4 Polyclonal Antibody

Catalog No. 720430
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-043-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-043-0 Supplier Invitrogen™ Supplier No. 720430

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

This antibody is predicted to react with Monkey, Horse, Cat, Pig.

This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is preferentially expressed in the skeletal muscle, and binds to the M-CAT regulatory element found in promoters of muscle-specific genes to direct their gene expression. Alternatively spliced transcripts encoding distinct isoforms, some of which are translated through the use of a non-AUG (UUG) initiation codon, have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEAD4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4
Gene Tead4
Gene Accession No. Q15561
Gene Alias EFTR-2; Etfr2; ETFR-2; ETF-related factor 2; ETF-related factor-2; FGF-regulated 19; FR-19; hRTEF-1B; related transcription enhancer factor 1B; RTEF1; RTEF-1; TCF13L1; Tcf13r1; TEA domain family member 4; TEA domain transcription factor 4; TEAD4; TEAD-4; TEF-1-related factor 1; TEF-1-related factor FR-19; TEF3; TEF-3; Tefr; Tefr1; TEFR-1; Tefr1a; Transcription factor 13-like 1; transcription factor RTEF-1; transcriptional enhancer factor 1-related; transcriptional enhancer factor 3; transcriptional enhancer factor TEF-3
Gene Symbols Tead4
Host Species Rabbit
Immunogen Protein corresponding to human TEAD4 [aa151-aa250].
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7004
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.