Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Teashirt homolog 1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282038AF488
This item is not returnable.
View return policy
Description
Teashirt homolog 1 Polyclonal antibody specifically detects Teashirt homolog 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Teashirt homolog 1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
10194 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunofluorescence | |
Alexa Fluor 488 | |
Antigen NY-CO-33, NY-CO-33, SDCCAG33, Serologically defined colon cancer antigen 33teashirt family zinc finger 1, teashirt homolog 1, teashirt zinc finger homeobox 1, TSH1teashirt zinc finger 1 | |
Teashirt homolog 1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human Teashirt homolog 1. The immunogen is located within amino acids 70 - 120 of Teashirt homolog 1. Amino Acid Squence: AHFKGSSSREEKEDPQ | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction