Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518868
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518868 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518868 Supplier Invitrogen™ Supplier No. PA518868
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

MARCH6 is an E3 ubiquitin ligase that localizes to endoplasmic reticulum membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEB4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MARCHF6
Gene Accession No. O60337
Gene Alias 3830408G03; 6-Mar; DOA10; doa10 homolog; E3 ubiquitin-protein ligase MARCH6; F830029L24Rik; KIAA0597; MARCH6; MARCH-VI; membrane associated ring finger 6; membrane associated ring-CH-type finger 6; membrane-associated ring finger (C3HC4) 6; membrane-associated ring finger (C3HC4) 6, E3 ubiquitin protein ligase; membrane-associated RING finger protein 6; membrane-associated RING-CH finger protein 6; membrane-associated RING-CH protein VI; mKIAA0597; Protein TEB-4; RING finger protein 176; RING-type E3 ubiquitin transferase MARCH6; RNF176; TEB4
Gene Symbols MARCHF6
Host Species Goat
Immunogen Synthetic peptide sequence (QGRRVIFQKVKE) corresponding to the internal amino acids of 38777 (aa 701-713).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10299
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.