Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565278 Supplier Invitrogen™ Supplier No. PA565278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HTRQWLKGLVR AWTVTAGYLL DLHSYLLGDQ EENENSANQQ VNNNQHARNN NAIPVVGEGL HAAHQAILQQ GGPVGFQPYR RPLN Highest antigen sequence identity to the following orthologs - mouse 95%, rat 94%.

MARCH6 is an E3 ubiquitin ligase that localizes to endoplasmic reticulum membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEB4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MARCHF6
Gene Accession No. O60337
Gene Alias 3830408G03; 6-Mar; DOA10; doa10 homolog; E3 ubiquitin-protein ligase MARCH6; F830029L24Rik; KIAA0597; MARCH6; MARCH-VI; membrane associated ring finger 6; membrane associated ring-CH-type finger 6; membrane-associated ring finger (C3HC4) 6; membrane-associated ring finger (C3HC4) 6, E3 ubiquitin protein ligase; membrane-associated RING finger protein 6; membrane-associated RING-CH finger protein 6; membrane-associated RING-CH protein VI; mKIAA0597; Protein TEB-4; RING finger protein 176; RING-type E3 ubiquitin transferase MARCH6; RNF176; TEB4
Gene Symbols MARCHF6
Host Species Rabbit
Immunogen Recombinant Human TEB4. Recombinant protein control fragment (Product #RP-105994).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10299
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.