Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEC Monoclonal Antibody (3A5)
GREENER_CHOICE

Catalog No. PIMA515546
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515546 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515546 Supplier Invitrogen™ Supplier No. MA515546
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15546 targets TEC in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15546 immunogen is purified recombinant fragment of TEC expressed in E. Coli. MA5-15546 detects TEC which has a predicted molecular weight of approximately 74kDa.

TEC is a non-receptor protein-tyrosine kinase containing a pleckstrin homology domain and is highly expressed in many hematopoietic cell lines. They are also key players in the regulation of the immune functions. Tec kinase is an integral component of T cell signaling and has a distinct role in T cell activation. This gene may be associated with myelodysplastic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEC
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3A5
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene Tec
Gene Accession No. P42680
Gene Alias cytoplasmic tyrosine kinase, Dscr28C related; MGC126760; MGC126762; PSCTK4; TEC; tec protein tyrosine kinase; tyrosine-protein kinase Tec
Gene Symbols Tec
Host Species Mouse
Immunogen Purified recombinant fragment of TEC expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100124696, 7006
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.