Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116493
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116493 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116493 Supplier Invitrogen™ Supplier No. PA5116493
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, Mouse liver, Mouse thymus, Mouse spleen, Rat liver Immunogen sequence: MNFNTILEEI LIKRSQQKKK TSPLNYKERL FVLTKSMLTY YEGRAEKKYR KGFIDVSKIK CVEIVKNDDG VIPCQNKYPF QVVHDANTLY IFAPSPQSRD LWVKKLKEEI KNNNNIMIKY HPKFWTDGSY QCCRQTEKLA PGCEKYNLFE SSIRKALPPA PETKKRRPPP PIPLEEEDNS EEIVVAMYDF QAAEGHDLRL ERGQEYLILE KNDVHWWRAR DKYGNEGYIP SNYVTGKKSN NLDQYEWYCR NMNRSKAEQL LRSEDKEGGF MVRDSSQPGL.

TEC is a non-receptor protein-tyrosine kinase containing a pleckstrin homology domain and is highly expressed in many hematopoietic cell lines. They are also key players in the regulation of the immune functions. Tec kinase is an integral component of T cell signaling and has a distinct role in T cell activation. This gene may be associated with myelodysplastic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEC
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Tec
Gene Accession No. P24604, P42680
Gene Alias cytoplasmic tyrosine kinase, Dscr28C related; MGC126760; MGC126762; PSCTK4; TEC; tec protein tyrosine kinase; tyrosine-protein kinase Tec
Gene Symbols Tec
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human Tec (NP_0032062).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100124696, 21682, 7006, 84492
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.