Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TEC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580101
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HEL whole cell, rat kidney tissue. IHC: Human Intestinal Cancer Tissue, Rat Liver Tissue.
TEC is a non-receptor protein-tyrosine kinase containing a pleckstrin homology domain and is highly expressed in many hematopoietic cell lines. They are also key players in the regulation of the immune functions. Tec kinase is an integral component of T cell signaling and has a distinct role in T cell activation. This gene may be associated with myelodysplastic syndrome.
Specifications
TEC | |
Polyclonal | |
Unconjugated | |
Tec | |
cytoplasmic tyrosine kinase, Dscr28C related; MGC126760; MGC126762; PSCTK4; TEC; tec protein tyrosine kinase; tyrosine-protein kinase Tec | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100124696, 7006, 84492 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.01mg sodium azide, 0.01mg thimerosal | |
P42680 | |
Tec | |
A synthetic peptide corresponding to a sequence at the C-terminus of human Tec (612-631aa FEDLLRTIDELVECEETFGR). | |
100 μg | |
Primary | |
Human, Rat, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction