Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TEC Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110697
Description
Immunogen sequence: SNYVTGKKSN NLDQYEWYCR NMNRSKAEQL LRSEDKEGGF MVRDSSQPGL YTVSLYTKFG GEGSSGFRHY HIKETTTSPK KYYLAEKHAF GSIPEIIEYH KHNAAGLVTR LRYPVSVKGK NAPTTAGFSY EKWEINPSEL TFMRELGSGL.
TEC is a non-receptor protein-tyrosine kinase containing a pleckstrin homology domain and is highly expressed in many hematopoietic cell lines. They are also key players in the regulation of the immune functions. Tec kinase is an integral component of T cell signaling and has a distinct role in T cell activation. This gene may be associated with myelodysplastic syndrome.
Specifications
| TEC | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P42680 | |
| Tec | |
| Recombinant Protein corresonding to Human TEC. Recombinant protein control fragment (Product #RP-95919). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Tec | |
| cytoplasmic tyrosine kinase, Dscr28C related; MGC126760; MGC126762; PSCTK4; TEC; tec protein tyrosine kinase; tyrosine-protein kinase Tec | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100124696, 7006 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction