Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex TECAN Extractor Rack Stabilizer
SDP

Catalog No. p-7222303
Click to view available options
Positions:
1
2
3

Holds and stabilizes sample tube racks for Tecan or other related extraction/processing machines.

  • Comes in 1, 2, or 3 rack capacity
  • Super durable and cleanable (alcohol resistant) PETG construction

Specifications

Product Type Rack
Autoclavable Not Autoclavable
Color Clear
Material Nylon-coated Stainless Steel
Positions 1
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Extractor Racks
Size Small
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.