Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TECPR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313113
This item is not returnable.
View return policy
Description
TECPR1 Polyclonal antibody specifically detects TECPR1 in Mouse samples. It is validated for Western BlotSpecifications
| TECPR1 | |
| Polyclonal | |
| Western Blot 1:500-1:3000 | |
| DKFZP434B0335, FLJ23419, FLJ90593, KIAA1358, tectonin beta-propeller repeat containing 1, tectonin beta-propeller repeat-containing protein 1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human TECPR1. The exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 25851 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction