Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TECPR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546637
Description
Peptide sequence: YTKDKKWNSC VRRRKWIRYR RYKSRDIWAK IPSKDDPKEL PDPFNDLSVG Sequence homology: Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
This gene encodes a tethering factor involved in autophagy. The encoded protein is found at autolysosomes, and is involved in targeting protein aggregates, damaged mitochondria, and bacterial pathogens for autophagy.
Specifications
TECPR1 | |
Polyclonal | |
Unconjugated | |
TECPR1 | |
2210010N04Rik; KIAA1358; LOW QUALITY PROTEIN: tectonin beta-propeller repeat-containing protein 1; mKIAA1358; RGD1306873; TECPR1; tectonin beta-propeller repeat containing 1; tectonin beta-propeller repeat-containing protein 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25851 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7Z6L1 | |
TECPR1 | |
Synthetic peptide directed towards the N-terminal of human TECPR1 (aa 148-197). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction