Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TECPR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119614
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
This gene encodes a tethering factor involved in autophagy. The encoded protein is found at autolysosomes, and is involved in targeting protein aggregates, damaged mitochondria, and bacterial pathogens for autophagy.
Specifications
TECPR1 | |
Polyclonal | |
Unconjugated | |
TECPR1 | |
2210010N04Rik; KIAA1358; LOW QUALITY PROTEIN: tectonin beta-propeller repeat-containing protein 1; mKIAA1358; RGD1306873; TECPR1; tectonin beta-propeller repeat containing 1; tectonin beta-propeller repeat-containing protein 1 | |
Rabbit | |
Protein A | |
RUO | |
25851, 70381 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.47 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q7Z6L1, Q80VP0 | |
TECPR1 | |
Recombinant protein encompassing a sequence within the N-terminus region of human TECPR1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction