Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TECR Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501570631
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0631 Supplier Bethyl Laboratories, Inc Supplier No. A305515A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine

Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. GPSN2 catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA. Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA (Moon and Horton, 2003 [PubMed 12482854]).[supplied by OMIM].

Specifications

Antigen TECR
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TECR
Gene Accession No. Q9CY27, Q9NZ01
Gene Alias GPSN2, MRT14, SC2, TER, glycoprotein, synaptic 2, synaptic glycoprotein SC2
Gene Symbols TECR
Host Species Rabbit
Immunogen Between 200 and 250
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106529, 9524
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.