Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tecrl Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569327
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569327 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569327 Supplier Invitrogen™ Supplier No. PA569327
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 83%; Horse: 85%; Human: 100%; Mouse: 100%; Rabbit: 85%; Rat: 100%.

The protein encoded by this gene contains a ubiquitin-like domain in the N-terminal region, three transmembrane segments and a C-terminal 3-oxo-5-alpha steroid 4-dehydrogenase domain. The protein belongs to the steroid 5-alpha reductase family. Mutations in this gene result in ventricular tachycardia, catecholaminergic polymorphic, 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tecrl
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TECRL
Gene Accession No. Q8BFZ1
Gene Alias D330017N19Rik; glycoprotein, synaptic 2-like; GPSN2L; SRD5A2L2; steroid 5 alpha-reductase 2-like 2; steroid 5-alpha-reductase 2-like 2 protein; synaptic glycoprotein SC2-like; TECRL; TERL; trans-2,3-enoyl-CoA reductase like; trans-2,3-enoyl-CoA reductase-like; Unknown (protein for MGC:128084)
Gene Symbols TECRL
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of mouse Tecrl.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243078
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.