Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TECTA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580102
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat testis tissue, mouse HEPA1-6 whole cell, human HepG2 whole cell. IHC: human intetsinal cancer tissue, human lung cancer tissue, human testis tissue.
Alpha-tectorin is a protein that in humans is encoded by the TECTA gene. The tectorial membrane is an extracellular matrix of the inner ear that contacts the stereocilia bundles of specialized sensory hair cells. Sound induces movement of these hair cells relative to the tectorial membrane, deflects the stereocilia, and leads to fluctuations in hair-cell membrane potential, transducing sound into electrical signals. Alpha-tectorin is one of the major noncollagenous components of the tectorial membrane. Mutations in the TECTA gene have been shown to be responsible for autosomal dominant nonsyndromic hearing impairment and a recessive form of sensorineural pre-lingual non-syndromic deafness.
Specifications
TECTA | |
Polyclonal | |
Unconjugated | |
Tecta | |
[a]-tectorin; alpha-tectorin; DFNA12; DFNA8; DFNB21; Tctna; TECTA; tectorin alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21683, 300653, 7007 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
O08523, O75443 | |
Tecta | |
A synthetic peptide corresponding to a sequence at the N-terminus of human TECTA (93-134aa RAFVAPFWADVHNGIRGEIYYRETMEPAILKRATKDIRKYFK). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction