Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TECTA Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA553868
Description
Immunogen sequence: VLHTFDGASY AFPSEFSYTL LKTCPERPEY LEIDINKKKP DAGPAWLRGL RILVADQEVK IGGIGASEVK LNGQE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

Specifications
TECTA | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75443 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7007 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Unconjugated | |
TECTA | |
TECTA | |
Recombit protein corresponding to Human TECTA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction