Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TECTB Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA523785

Catalog No. PIPA523785


Only null left
Add to Cart

Description

Description

TECTB Polyclonal Antibody for Western Blot, IHC (P)

The genes for alpha-tectorin (MIM 602574) and beta-tectorin encode the major noncollagenous proteins of the tectorial membrane of the cochlea.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TECTB
Polyclonal
Unconjugated
TECTB
[b]-tectorin; beta-tectorin; Tctnb; Tectb; tectorin beta
Rabbit
Antigen affinity chromatography, Protein A
RUO
21684, 6975
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
O08524, Q96PL2
TECTB
KLH conjugated synthetic peptide between 54-83 amino acids from the N-terminal region of human TECTB.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.