Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEDC1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298663AF532
Description
TEDC1 Polyclonal specifically detects TEDC1 in Human samples. It is validated for Western Blot.Specifications
TEDC1 | |
Polyclonal | |
Western Blot | |
C14orf80, chromosome 14 open reading frame 80 | |
Produced in rabbits immunized with E. coli-derived Human TEDC1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
283643 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction