Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TEF Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106495
Description
Antibody detects endogenous levels of total TEF.
TEF (thyrotrophic embryonic factor), also known as KIAA1655, is a 303 amino acid nuclear transcription factor that belongs to the bZIP (basic region/leucine zipper) family and PAR (proline and acidic amino acid-rich) subfamily. TEF binds DNA as either a homodimer or heterodimer, and is known to transactivate the TSH beta promoter. While broadly expressed in adults, TEF is only found in developing embryonic anterior pituitary gland. TEF accumulates according to a robust circadian rhythm and has also been found to inhibit cell growth by downregulating beta chain expression of cytokine receptors. The functional domains of TEF are highly homologous with other members of the PAR-bZIP subfamily, including albumin D box-binding protein (DABP), human hepatic leukemia factor (HLF) and chicken vitellogenin gene-binding protein (VBP).
Specifications
| TEF | |
| Polyclonal | |
| Unconjugated | |
| TEF | |
| 2310028D20Rik; KIAA1655; TEF; TEF, PAR bZIP transcription factor; thyrotroph embryonic factor; thyrotrophic embryonic factor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21685, 29362, 7008 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P41224, Q10587, Q9JLC6 | |
| TEF | |
| A synthesized peptide derived from human TEF(Accession Q10587), corresponding to amino acid residues R204-R254. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction