Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536187
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536187 Supplier Invitrogen™ Supplier No. PA536187
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 33 kDa. Purity is >95% by SDS-PAGE.

TEF (thyrotrophic embryonic factor), also known as KIAA1655, is a 303 amino acid nuclear transcription factor that belongs to the bZIP (basic region/leucine zipper) family and PAR (proline and acidic amino acid-rich) subfamily. TEF binds DNA as either a homodimer or heterodimer, and is known to transactivate the TSH beta promoter. While broadly expressed in adults, TEF is only found in developing embryonic anterior pituitary gland. TEF accumulates according to a robust circadian rhythm and has also been found to inhibit cell growth by downregulating beta chain expression of cytokine receptors. The functional domains of TEF are highly homologous with other members of the PAR-bZIP subfamily, including albumin D box-binding protein (DABP), human hepatic leukemia factor (HLF) and chicken vitellogenin gene-binding protein (VBP).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEF
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene TEF
Gene Accession No. P41224, Q10587, Q9JLC6
Gene Alias 2310028D20Rik; KIAA1655; TEF; TEF, PAR bZIP transcription factor; thyrotroph embryonic factor; thyrotrophic embryonic factor
Gene Symbols TEF
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 200-250 of human TEF.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21685, 29362, 7008
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.