Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEF1 Rabbit anti-Human, Clone: 3Q10Q8, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
| Antigen | TEF1 |
|---|---|
| Clone | 3Q10Q8 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
TEF1 Monoclonal antibody specifically detects TEF1 in Human samples. It is validated for Western BlotSpecifications
| TEF1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, Cell Biology | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 7003 | |
| IgG | |
| Affinity purified |
| 3Q10Q8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| NTEF-1, protein GT-IIC, TEA domain family member 1 (SV40 transcriptional enhancer factor), TEAD-1 | |
| A synthesized peptide derived from human TEF1 | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title