Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEF1 Rabbit anti-Human, Mouse, Rat, Clone: 2E7N10, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | TEF1 |
---|---|
Clone | 2E7N10 |
Applications | ChIP Assay, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
TEF1 Monoclonal antibody specifically detects TEF1 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, ImmunoprecipitationSpecifications
TEF1 | |
ChIP Assay, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cardiovascular Biology, Cell Biology | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7003 | |
IgG | |
Affinity purified |
2E7N10 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
NTEF-1, protein GT-IIC, TEA domain family member 1 (SV40 transcriptional enhancer factor), TEAD-1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TEF1 (P28347). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title