Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEF3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989191
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9191 Supplier Bioss Supplier No. BS9603R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is preferentially expressed in the skeletal muscle, and binds to the M-CAT regulatory element found in promoters of muscle-specific genes to direct their gene expression. Alternatively spliced transcripts encoding distinct isoforms, some of which are translated through the use of a non-AUG (UUG) initiation codon, have been described for this gene.

Specifications

Antigen TEF3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Tead4
Gene Accession No. Q15561
Gene Alias BOS_6026; EFTR-2; Etfr2; ETFR-2; ETF-related factor 2; ETF-related factor-2; FGF-regulated 19; FR-19; hRTEF-1B; LOC100341523; related transcription enhancer factor 1B; RTEF1; RTEF-1; TCF13L1; Tcf13r1; TEA domain family member 4; TEA domain family member 4-like; TEA domain transcription factor 4; Tead4; TEAD-4; TEF-1-related factor 1; TEF-1-related factor FR-19; Tef3; TEF-3; Tefr; Tefr1; TEFR-1; Tefr1a; Transcription factor 13-like 1; Transcription factor RTEF-1; transcriptional enhancer factor 1-related; transcriptional enhancer factor 3; Transcriptional enhancer factor TEF-3
Gene Symbols Tead4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TEF3/TEAD4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7004
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.