Learn More
Invitrogen™ TEF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121103
Description
Positive test controls include: Mouse heart, Mouse lung, Rat heart. The target is usually found in the following locations: Nucleus. Immunogen sequence: SSAQIVSASV LQNKFSPPSP LPQAVFSTSS RFWSSPPLLG QQPGPSQDIK PFAQPAYPIQ PPLPPTLSSY EPLAPLPSAA ASVPVWQDRT IASSRLRLLE YSAFMEVQRD PDTYSKHLFV HIGQTNPAFS DPPLEAVDVR QIYDKFPEKK GGLKELYEKG PPNAFFLVKF WADLNSTIQE GPGAFYGVSS QYSSADSMTI S.
This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is predominantly expressed in the placenta and is involved in the transactivation of the chorionic somatomammotropin-B gene enhancer. Translation of this protein is initiated at a non-AUG (AUA) start codon.
Specifications
TEF5 | |
Polyclonal | |
Unconjugated | |
TEAD3 | |
DTEF-1; ETFR-1; ETF-related factor 1; Tcf13r2; TEA domain family member 3; TEA domain family member 5; TEA domain transcription factor 3; Tead3; TEAD-3; TEAD5; TEF5; TEF-5; transcriptional enhancer factor 5; transcriptional enhancer factor TEF-5; transcriptional enhancer factor TEF-5 (DTEF-1) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
21678, 294299, 7005 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.62 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70210, Q99594 | |
TEAD3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 130-330 of human TEAD3 (NP_003205.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.