Learn More
Invitrogen™ TEFM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597096
Description
Immunogen sequence: KSTTPKKITP NVTFCDENAK EPENALDKLF SSEQQASILH VLNTASTKEL EAFRLLRGRR SINIVEHREN FGPFQNLESL MNVPLFKYKS TVQVCNSILC PKTGREKRKS PENRFLRKLL KPDIERERLK AVNSIISIVF GTRRIAWAHL DRKLTVLDWQ QSDRWSLMRG IYSSS; Positive Samples: HL-60, K-562, Mouse liver, Rat liver; Cellular Location: Mitochondrion matrix, mitochondrion nucleoid.
TEFM is a transcription elongation factor which increases mitochondrial RNA polymerase processivity. It regulates transcription of the mitochondrial genome, including genes important for the oxidative phosphorylation machinery.
Specifications
TEFM | |
Polyclonal | |
Unconjugated | |
TEFM | |
1110002N22Rik; C17orf42; C9H17orf42; RGD1306595; Tefm; transcription elongation factor of mitochondria; transcription elongation factor, mitochondrial | |
Rabbit | |
Affinity Chromatography | |
RUO | |
287554, 68550, 79736 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.75 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4KM51, Q5SSK3, Q96QE5 | |
TEFM | |
Recombinant fusion protein containing a sequence corresponding to amino acids 36-210 of human TEFM (NP_0789593). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.