Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEFM Polyclonal Antibody, Invitrogen™

Catalog No. PIPA554887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554887 Supplier Thermo Scientific Supplier No. PA554887

Rabbit Polyclonal Antibody

Immunogen sequence: RRSINIVEHR ENFGPFQNLE SLMNVPLFKY KSTVQVCNSI LCPKTGREKR KSPENRFLRK LLKPDIERER LKAVNSIISI V Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 65%.

TEFM is a transcription elongation factor which increases mitochondrial RNA polymerase processivity. It regulates transcription of the mitochondrial genome, including genes important for the oxidative phosphorylation machinery.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEFM
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TEFM
Gene Accession No. Q96QE5
Gene Alias 1110002N22Rik; C17orf42; C9H17orf42; RGD1306595; Tefm; transcription elongation factor of mitochondria; transcription elongation factor, mitochondrial
Gene Symbols TEFM
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TEFM
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79736
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.