Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEFM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546121
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546121 Supplier Invitrogen™ Supplier No. PA546121
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RSSLYWALHN FCCRKKSTTP KKITPNVTFC DENAKEPENA LDKLFSSEQQ Sequence homology: Dog: 77%; Human: 100%; Pig: 79%.

TEFM is a transcription elongation factor which increases mitochondrial RNA polymerase processivity. It regulates transcription of the mitochondrial genome, including genes important for the oxidative phosphorylation machinery.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEFM
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TEFM
Gene Accession No. Q96QE5
Gene Alias 1110002N22Rik; C17orf42; C9H17orf42; RGD1306595; Tefm; transcription elongation factor of mitochondria; transcription elongation factor, mitochondrial
Gene Symbols TEFM
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TEFM (aa 21-70).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79736
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.