Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Teicoplanin, G-Biosciences
SCI_ED

Catalog No. S139787
Click to view available options
Quantity:
100 mg
1000 mg

Teicoplanin is a glycopeptide antibiotic isolated from Actinoplanes teichomyceticus. It is a mixture of several teicoplanins.

Teicoplanin is effective against gram-positive bacteria resistant to Beta-lactam antibiotics. It has been effective against methicillin-resistant Staphylococcus aureus and Enterococcus faecalis.

Chemical Identifiers

Molecular Weight (g/mol) 1879.66 g/mol

Specifications

Quantity 100 mg
Percent Purity >95%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.