Learn More
TEIF Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302653A

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a transcriptional regulator belonging to the SCY1-like family of kinase-like proteins. The protein has a divergent N-terminal kinase domain that is thought to be catalytically inactive, and can bind specific DNA sequences through its C-terminal domain. It activates transcription of the telomerase reverse transcriptase and DNA polymerase beta genes. The protein has been localized to the nucleus, and also to the cytoplasm and centrosomes during mitosis. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
TEIF | |
Polyclonal | |
Unconjugated | |
SCYL1 | |
GKLP, NKTL, NTKL, P105, TAPK, TEIF, TRAP, N-terminal kinase-like protein, SCY1-like protein 1, coated vesicle-associated kinase of 90 kDa, likely ortholog of mouse N-terminal kinase-like protein, telomerase regulation-associated protein, telomerase transcriptional element-interacting factor, telomerase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57410, 78891 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q96KG9, Q9EQC5 | |
Scyl1 | |
Between 758 and 808 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.