Learn More
Invitrogen™ TEK Recombinant Rabbit Monoclonal Antibody (116), PE

Description
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human TIE2 (TEK) (Met1-Lys745) and conjugated with PE under optimum conditions, the unreacted PE was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.
Specifications
Specifications
| Antigen | TEK |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | 116 |
| Concentration | 100 μg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.5% BSA and 0.03% ProClin 300 |
| Gene | TEK |
| Gene Accession No. | Q02763 |
| Gene Alias | AA517024; Angiopoietin-1 receptor; CD202B; CD202b antigen; EC 2.7.10; EC 2.7.10.1; Endothelial tyrosine kinase; endothelial-specific receptor tyrosine kinase; endothelium-specific receptor tyrosine kinase 2; endothelium-specific receptor tyrosine kinase Tie-2; hTIE2; Hyk; mTIE2; p140 TEK; protein-tyrosine kinase Tie2; STK1; Tek; TEK receptor tyrosine kinase; TEK tyrosine kinase, endothelial; TEKT1; Tektin-1; Tie2; tie-2; tunica interna endothelial cell kinase; Tyrosine kinase with Ig and EGF homology domains-2; tyrosine kinases that contain immunoglobulin-like loops and epidermal growth factor-similar domains 2; tyrosine-protein kinase receptor TEK; Tyrosine-protein kinase receptor Tie-2; VMCM; VMCM1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.