Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEKT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590307
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590307 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590307 Supplier Invitrogen™ Supplier No. PA590307
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAKLLQPPPK FLPSEWHIAN KNQYHRADAQ RSRSERLVAE SQRLVDEIEK TTRKSQSDVN KKLEQRLEEV QFWKKELDDK LEQLVNVTDD LLIYKIRLEK ALETLKEPLH ITETCLAYRE KRIGIDLVHD TVEHELIKEA EIIQGIMALL TRTLEEASEQ IRMNRSAKYN LEKDLKDKFV ALTIDDICFS LNNNSPNIRY SENAVRIEPN SVSLEDWLDF SSTNVEKADK QRNNSLMLKA LVDRILSQTA NDLRKQCDVV DTAFKNGLKD TKDARDKLAD HLAKVMEEIA SQEKNITALE; Positive Samples: Mouse testis, Rat testis; Cellular Location: Cytoplasm, cilium axoneme, cytoskeleton, flagellum axoneme.

This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. This gene is predominantly expressed in the testis and in mouse, tektin 1 mRNA was localized to the spermatocytes and round spermatids in the seminiferous tubules, indicating that it may play a role in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEKT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TEKT1
Gene Accession No. Q99JD2, Q9DAJ2
Gene Alias MT14; TEKT1; tektin 1; tektin-1
Gene Symbols TEKT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TEKT1 (NP_4445151).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21689, 85270
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.