Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEKT2 Monoclonal Antibody (OTI1E11)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527238

Catalog No. PIMA527238


Only null left
Add to Cart

Description

Description

TEKT2 Monoclonal Antibody for Western Blot

Structural component of ciliary and flagellar microtubules. Plays a key role in the assembly or attachment of the inner dynein arm to microtubules in sperm flagella and tracheal cilia. Forms filamentous polymers in the walls of ciliary and flagellar microtubules.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TEKT2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UIF3
TEKT2
Human recombinant protein fragment corresponding to amino acids 1-244 of TEKT2 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI1E11
Unconjugated
TEKT2
AU014974; h-tektin-t; TEKT2; TEKTB1; tektin 2; tektin 2 (testicular); tektin-2; tektin-B1; TEKTIN-T; Testicular tektin; testicular tektin B1-like protein
Mouse
Affinity Chromatography
RUO
27285
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.