Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEKT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | TEKT2 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8805
![]()
|
Proteintech Group Inc
135181AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Structural component of ciliary and flagellar microtubules. Plays a key role in the assembly or attachment of the inner dynein arm to microtubules in sperm flagella and tracheal cilia. Forms filamentous polymers in the walls of ciliary and flagellar microtubules.Specifications
| TEKT2 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q6AYM2, Q922G7, Q9UIF3 | |
| 24084, 27285, 298532 | |
| TEKT2 Fusion Protein Ag4457 | |
| Primary | |
| -20°C | |
| TEKT2 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| h tektin t, TEKT2, TEKTB1, Tektin 2, tektin 2 (testicular), Tektin B1, TEKTIN T, Testicular tektin | |
| Tekt2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title