Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEKT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144737 Supplier Invitrogen™ Supplier No. PA5144737
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: REKRMGIDLV HDEVEAQLLT EVDTILCCQE RMKLHLDKAI AQLAANRASQ HELEKDLSDK QTAYRIDDKC HHLRNTSDGV GYFRGVERVD ATVS Antigen sequence identity: Mouse 83%; Rat 82%.

This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. The exact function of this gene is not known. Structural component of ciliary and flagellar microtubules. Forms filamentous polymers in the walls of ciliary and flagellar microtubules. Required for progressive sperm mobility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEKT3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TEKT3
Gene Accession No. Q9BXF9
Gene Alias 4933407G07Rik; OMC45; TEKT3; tektin 3; tektin-3; testicular microtubules-related protein
Gene Symbols TEKT3
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 202-295. Recombinant protein control fragment (Product #RP-110065).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64518
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.