Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TEKT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560687
Description
Immunogen sequence: RVGCTLTTTY AHPRPTPTNF LPAISTMASS YRDRFPHSNL THSLSLPWRP STYYKVASNS PSVAPYCTRS QRVSENTMLP F Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 70%.
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. The exact function of this gene is not known. Structural component of ciliary and flagellar microtubules. Forms filamentous polymers in the walls of ciliary and flagellar microtubules. Required for progressive sperm mobility.
Specifications
TEKT3 | |
Polyclonal | |
Unconjugated | |
TEKT3 | |
4933407G07Rik; OMC45; TEKT3; tektin 3; tektin-3; testicular microtubules-related protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64518 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BXF9 | |
TEKT3 | |
Recombinant protein corresponding to Human TEKT3. Recombinant protein control fragment (Product #RP-93244). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction