Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEKT3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | TEKT3 |
---|---|
Dilution | Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TEKT3 Polyclonal antibody specifically detects TEKT3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TEKT3 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
FLJ32828, tektin 3, tektin-3, testicular microtubules-related protein | |
This antibody was developed against Recombinant Protein corresponding to amino acids: WYRSNLTNYQESNTSRHNSEKLRVDTSRLIQDKYQQTRKTQADTTQNLGERVNDIGFWKSEIIHELDEMIGETNALTDVKKRLERALMETEAP | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
64518 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title