Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEKT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531587 Supplier Invitrogen™ Supplier No. PA531587
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TEKT4 is structural component of ciliary and flagellar microtubules. TEKT4 forms filamentous polymers in the walls of ciliary and flagellar microtubules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEKT4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene TEKT4
Gene Accession No. Q8WW24
Gene Alias 1700010L19Rik; RGD1308075; Tek4; Tekt4; tektin 4; Tektin4; tektin-4; Testicular microtubules-related protein 4
Gene Symbols TEKT4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Tektin 4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150483
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.