Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tellurium, 99+%, (trace metal basis), powder

Catalog No. p-221650
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle

CAS: 13494-80-9 | Te | 127.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13494-80-9
Molecular Formula Te
Molecular Weight (g/mol) 127.60
MDL Number MFCD00134062
InChI Key PORWMNRCUJJQNO-UHFFFAOYSA-N
Synonym tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
PubChem CID 6327182
ChEBI CHEBI:30452
IUPAC Name tellurium
SMILES [Te]

Specifications

Boiling Point 990.0°C
Melting Point 450.0°C
Color Black to Gray
Physical Form Powder
Quantity 10 g
Assay Percent Range 99+%
Linear Formula Te
Fieser 15,298
Merck Index 15, 9263
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in h2so4, hno3 and koh, insoluble in benzene and carbon disulfide
Formula Weight 127.6
Percent Purity ≥99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Tellurium, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
May cause an allergic skin reaction.
May cause long lasting harmful effects to aquatic life.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position
EINECSNumber 236-813-4
RTECSNumber WY2625000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.