Alfa Aesar™ Tellurium pieces, 99.9999% (metals basis), zone refined

Manufacturer: Alfa Aesar™ 1107609

Quantity 10g

 View more versions of this product

Catalog No. AA1107609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13494-80-9
    MDL Number MFCD00134062
    Chemical Name or Material Tellurium pieces, Zone refined

    Specifications

    Quantity 10g
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P304+P340-P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H301-H332
    Toxic if swallowed.
    Harmful if inhaled.
    Odor Odorless
    TSCA TSCA
    Physical Form Pieces
    MDL Number MFCD00134062
    CAS 13494-80-9
    Health Hazard 1 Danger
    Chemical Name or Material Tellurium pieces, Zone refined
    Assay 99.9999%, (metals basis)

    GHS H Statement
    H301-H332
    Toxic if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P301+P310-P304+P340-P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit