Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TELO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559583
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559583 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559583 Supplier Invitrogen™ Supplier No. PA559583
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKAVLICLAQ LGEPELRDSR DELLASMMAG VKCRLDSSLP PVRRLGMIVA EVVSARIHPE GPPLKFQYEE DELSLELLAL ASPQPAGDGA SEAGTSLVPA TA Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 75%.

TELO2 is a protein that functions as an S-phase checkpoint protein in the cell cycle. The protein may also play a role in DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TELO2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TELO2
Gene Accession No. Q9Y4R8
Gene Alias 1200003M09Rik; AI415602; CLK2; hCLK2; KIAA0683; mKIAA0683; protein clk-2 homolog; RGD1305553; Tel2; TEL2, telomere maintenance 2, homolog; TEL2, telomere maintenance 2, homolog (S. cerevisiae); Telo2; Telomere length regulation protein TEL2 homolog; telomere maintenance 2; YHFS
Gene Symbols TELO2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TELO2. Recombinant protein control fragment (Product #RP-98676).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9894
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.