Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TELO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116718 Supplier Invitrogen™ Supplier No. PA5116718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: K-562, HeLa Immunogen sequence: ASEAGTSLVP ATAEPPAETP AEIVDGGVPQ AQLAGSDSDL DSDDEFVPYD MSGDRELKSS KAPAYVRDCV EALTTSEDIE RWEAALRALE GLVYRSPTAT R.

TELO2 is a protein that functions as an S-phase checkpoint protein in the cell cycle. The protein may also play a role in DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TELO2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TELO2
Gene Accession No. Q9DC40, Q9Y4R8
Gene Alias 1200003M09Rik; AI415602; CLK2; hCLK2; KIAA0683; mKIAA0683; protein clk-2 homolog; RGD1305553; Tel2; TEL2, telomere maintenance 2, homolog; TEL2, telomere maintenance 2, homolog (S. cerevisiae); Telo2; Telomere length regulation protein TEL2 homolog; telomere maintenance 2; YHFS
Gene Symbols TELO2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 450-550 of human TELO2 (NP_0571952).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71718, 9894
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.