Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Telomerase Reverse Transcriptase (TERT) Monoclonal Antibody (2C4), Abbomax

Catalog No. 502204439
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4439 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4439 Supplier Abbomax Inc Supplier No. 606350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: HeLa whole cell lysate Cellular location: Nucleus.

Telomerase Reverse Transcriptase (TERT) Monoclonal antibody specifically detects Telomerase Reverse Transcriptase (TERT) in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Telomerase Reverse Transcriptase (TERT)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2C4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene TERT
Gene Accession No. O14746
Gene Alias CMM9; DKCA2; DKCB4; EST2; HEST2; hTERT; hTRT; PFBMFT1; RGD:70494}; TCS1; Telomerase catalytic subunit; telomerase reverse transcriptase; telomerase reverse transcriptase beta; telomerase-associated protein 2; TERT; tert {ECO:0000312; TP2; TR; TRT
Host Species Mouse
Immunogen A recombinant protein of human TERT
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7015
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.