Learn More
Cole-Parmer™ TELOS™ neo™ PAX SPE Columns
Mixed-mode strong anion exchange columns for acidic analyte extraction
Supplier: Cole-Parmer™ 0647431
Description
- High surface area and capacity ensure highly reliable retention characteristics
- No secondary interactions from silanol groups for accurate results
- Higher sample loading due to high surface area
- Recover clean extracts and shorten the time spent on method development using TELOS neo polymeric SPE columns.
- High surface area and capacity allow the use of smaller sorbent masses, leading to reduced elution volumes and evaporation/reconstitution steps.
- Polymeric sorbents are water-wettable and not affected by drying out, providing better flow characteristics, retention, and elution.
- Columns provide high and reproducible recoveries for a wide range of analytes, including acidic, neutral, basic, and multi-charged analytes.
Specifications
| Polymer | |
| Mixed-mode Strong Anion Exchange | |
| 60 μm | |
| 6 mL |
| 200 mg | |
| Conventional HPLC System | |
| Solid Phase Extraction Column |