Learn More
Cole-Parmer™ TELOS™ neo™ PAX SPE Columns
Description
- High surface area and capacity ensure highly reliable retention characteristics
- No secondary interactions from silanol groups for accurate results
- Higher sample loading due to high surface area
- Recover clean extracts and shorten the time spent on method development using TELOS neo polymeric SPE columns.
- High surface area and capacity allow the use of smaller sorbent masses, leading to reduced elution volumes and evaporation/reconstitution steps.
- Polymeric sorbents are water-wettable and not affected by drying out, providing better flow characteristics, retention, and elution.
- Columns provide high and reproducible recoveries for a wide range of analytes, including acidic, neutral, basic, and multi-charged analytes.
Specifications
Specifications
| Column Phase | Polymer |
| Bed Weight | 30 mg |
| For Use With (Application) | Mixed-mode Strong Anion Exchange |
| For Use With (Equipment) | Conventional HPLC System |
| Particle Size | 60 μm |
| Type | Solid Phase Extraction Column |
| Volume (Metric) | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.