Learn More
Cole-Parmer™ TELOS™ neo™ PCX SPE Columns
Mixed-mode strong cation exchange columns for basic analyte extraction
Supplier: Cole-Parmer™ 0647453
Description
- High surface area and capacity ensure highly reliable retention characteristics
- No secondary interactions from silanol groups for accurate results
- Higher sample loading due to high surface area
- Recover clean extracts and shorten the time spent on method development using TELOS neo PCX SPE columns.
- Mixed-mode sorbents exhibit a dual retention mechanism to significantly reduce the matrix components in the final extract.
- High surface area and capacity allow the use of smaller sorbent masses, leading to reduced elution volumes and evaporation/reconstitution steps.
- Polymeric sorbents are water-wettable and not affected by drying out, providing better flow characteristics, retention, and elution.
Specifications
Polymer | |
Mixed-mode Strong Cation Exchange | |
60 μm | |
3 mL |
60 mg | |
Conventional HPLC System | |
Solid Phase Extraction Column |