Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEM7 Monoclonal Antibody (13)
GREENER_CHOICE

Catalog No. PIMA531119
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531119 Supplier Invitrogen™ Supplier No. MA531119
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human TEM7/PLXDC1.

Recently, using SAGE (Serial Analysis of Gene Expression) technology, St. Croix et al, have identified 46 genes, whose expression is specifically elevated in tumor-associated endothelium. Nine of these genes were prominently expressed only in tumor endothelial cells (EC), but were absent or barely detectable in normal ECs, and named as Tumor Endothelial Markers (TEMs, TEM 1-9). TEM7 (Tumor endothelial marker 7) transcripts are specifically expressed in the endothelium of colorectal cancer, primary cancers of lung, pancreas, breast, and brain. TEM7 is expressed specifically in endothelium of these cancers, whether primary or metastasis. The other six members of this family (TEM1, 3, 4, 5, 8, and 9) also show similar expression pattern in lung and brain tumors, and liver metastasis. Since most of the genes expressed differentially in tumor endothelium are also expressed during angiogenesis, these newly discovered genes might provide important resources for basic and clinical studies of human angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEM7
Applications ELISA
Classification Monoclonal
Clone 13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PLXDC1
Gene Accession No. Q8IUK5
Gene Alias 2410003I07Rik; ADP-ribosylation factor-like 12; AI848450; Arl12; DKFZp686F0937; FLJ36270; FLJ45632; plexin domain containing 1; plexin domain-containing protein 1; PLXDC1; TEM3; TEM7; TEM7 related; tumor endothelial marker 3; Tumor endothelial marker 7; tumor endothelial marker7
Gene Symbols PLXDC1
Host Species Mouse
Immunogen Recombinant Human TEM7/PLXDC1 Protein (Met1-Thr426).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57125
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.